Assay Details
| Gene Symbol: KIR2DS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097567.1 | NP_001091036.1 | 5 - 6 | 936 | 117 | |
| GenBank mRNA | AY075103.1 | - | 5 - 6 | 932 | 117 | |
| EF197119.1 | - | 5 - 6 | 936 | 117 | ||
More Information
Set Membership: |
|

KIR2DS1
Genomic Map