Assay Details
| Gene Symbol: IFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174350.1 | NP_776775.1 | 1 | 147 | 83 | |
| GenBank mRNA | EU276065.1 | - | 1 | 95 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFNB1| Gene Symbol: IFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174350.1 | NP_776775.1 | 1 | 147 | 83 | |
| GenBank mRNA | EU276065.1 | - | 1 | 95 | 83 | |
Set Membership: |
|