Assay Details
| Gene Symbol: KIR3DL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181451.1 | NP_852116.1 | 8 - 9 | 1173 | 152 | |
| GenBank mRNA | AF490402.1 | - | 8 - 9 | 1173 | 152 | |
More Information
Set Membership: |
|

KIR3DL1
Genomic Map