Assay Details
| Gene Symbol: MRPL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034274.2 | NP_001029446.1 | 2 - 3 | 293 | 151 | |
| GenBank mRNA | BC102348.1 | - | 2 - 3 | 294 | 151 | |
More Information
Set Membership: |
|

MRPL17
Genomic Map