Assay Details
| Gene Symbol: KRT34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075951.1 | NP_001069419.1 | 3 - 4 | 640 | 61 | |
| GenBank mRNA | BC118281.1 | - | 3 - 4 | 640 | 61 | |
More Information
Set Membership: |
|

KRT34
Genomic Map