Assay Details
| Gene Symbol: APON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003907.1 | NP_001003907.1 | 1 | 947 | 131 | |
| GenBank mRNA | AY583019.1 | - | 1 | 947 | 131 | |
| Gene Symbol: APOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC112639.1 | - | 2 | 859 | 131 | |
More Information
Set Membership: |
|

APON
Genomic Map