Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046060.2 | NP_001039525.1 | 3 | 351 | 118 | |
| GenBank mRNA | BC114879.1 | - | 3 | 352 | 118 | |
More Information
Set Membership: |
|

GIMAP4
Genomic Map