Assay Details
| Gene Symbol: TUBB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046373.1 | NP_001039838.1 | 4 | 1552 | 113 | |
| GenBank mRNA | BC113260.1 | - | 4 | 1552 | 113 | |
More Information
Set Membership: |
|

TUBB6
Genomic Map