Assay Details
| Gene Symbol: MPEG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046464.1 | NP_001039929.1 | 1 | 3944 | 126 | |
| GenBank mRNA | BC105415.1 | - | 1 | 3944 | 126 | |
More Information
Set Membership: |
|

MPEG1
Genomic Map