Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075538.2 | NP_001069006.1 | 3 | 943 | 138 | |
| GenBank mRNA | BC123543.1 | - | 3 | 944 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB9A| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075538.2 | NP_001069006.1 | 3 | 943 | 138 | |
| GenBank mRNA | BC123543.1 | - | 3 | 944 | 138 | |
Set Membership: |
|