Assay Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076411.2 | NP_001069879.1 | 1 | 152 | 105 | |
| GenBank mRNA | BC111353.1 | - | 1 | 154 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPF3B| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076411.2 | NP_001069879.1 | 1 | 152 | 105 | |
| GenBank mRNA | BC111353.1 | - | 1 | 154 | 105 | |
Set Membership: |
|