Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078147.1 | NP_001071615.1 | 1 | 250 | 150 | |
| GenBank mRNA | BC113235.1 | - | 1 | 250 | 150 | |
More Information
Set Membership: |
|

DCAF16
Genomic Map