Assay Details
| Gene Symbol: H1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083425.2 | NP_001076894.1 | 1 | 1545 | 131 | |
| GenBank mRNA | BC133454.1 | - | 1 | 1547 | 131 | |
More Information
Set Membership: |
|

H1-2
Genomic Map