Assay Details
| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101102.1 | NP_001094572.1 | 5 | 2452 | 78 | |
| GenBank mRNA | BC151650.1 | - | 5 | 2452 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MDFIC| Gene Symbol: MDFIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101102.1 | NP_001094572.1 | 5 | 2452 | 78 | |
| GenBank mRNA | BC151650.1 | - | 5 | 2452 | 78 | |
Set Membership: |
|