Assay Details
| Gene Symbol: PGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102335.1 | NP_001095805.1 | 11 | 2953 | 116 | |
| GenBank mRNA | BC148864.1 | - | 11 | 2953 | 116 | |
More Information
Set Membership: |
|

PGM5
Genomic Map