Assay Details
| Gene Symbol: PSAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103267.1 | NP_001096737.1 | 1 | 2352 | 156 | |
| GenBank mRNA | BC149798.1 | - | 1 | 2352 | 156 | |
More Information
Set Membership: |
|

PSAPL1
Genomic Map