Assay Details
| Gene Symbol: POU3F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105451.2 | NP_001098921.1 | 1 | 2053 | 107 | |
| GenBank mRNA | BC153279.1 | - | 1 | 2054 | 107 | |
More Information
Set Membership: |
|

POU3F1
Genomic Map