Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105501.2 | NP_001098971.1 | 7 | 1152 | 124 | |
| GenBank mRNA | BC149886.1 | - | 7 | 1153 | 124 | |
More Information
Set Membership: |
|

UBFD1
Genomic Map