Assay Details
| Gene Symbol: RNASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105511.1 | NP_001098981.1 | 2 | 646 | 145 | |
| GenBank mRNA | BC142288.1 | - | 2 | 646 | 145 | |
More Information
Set Membership: |
|

RNASE2
Genomic Map