Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110774.1 | NP_001104244.1 | 1 | 53 | 75 | |
| GenBank mRNA | BC122600.1 | - | 1 | 53 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122600.1, NM_001110774.1 transcripts.
Set Membership: |
|

SPTSSB
Genomic Map