Assay Details
| Gene Symbol: CHRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174270.2 | NP_776695.1 | 4 | 2248 | 118 | |
| GenBank mRNA | U08286.1 | - | 4 | 2248 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRM3| Gene Symbol: CHRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174270.2 | NP_776695.1 | 4 | 2248 | 118 | |
| GenBank mRNA | U08286.1 | - | 4 | 2248 | 118 | |
Set Membership: |
|