Assay Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176675.3 | NP_788848.1 | 1 | 441 | 151 | |
| GenBank mRNA | BC102366.1 | - | 1 | 441 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COX6B1| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176675.3 | NP_788848.1 | 1 | 441 | 151 | |
| GenBank mRNA | BC102366.1 | - | 1 | 441 | 151 | |
Set Membership: |
|