Assay Details
| Gene Symbol: KIR3DL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098089.1 | NP_001091558.1 | 5 | 745 | 69 | |
| GenBank mRNA | EF197118.1 | - | 5 | 745 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIR3DL2| Gene Symbol: KIR3DL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098089.1 | NP_001091558.1 | 5 | 745 | 69 | |
| GenBank mRNA | EF197118.1 | - | 5 | 745 | 69 | |
Set Membership: |
|