Assay Details
| Gene Symbol: PCGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144106.1 | NP_001137578.1 | 7 - 8 | 584 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCGF2| Gene Symbol: PCGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144106.1 | NP_001137578.1 | 7 - 8 | 584 | 69 | |
Set Membership: |
|