Assay Details
| Gene Symbol: GLYATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145061.1 | NP_001138533.1 | 1 - 2 | 95 | 100 | |
| GenBank mRNA | DT841858.1 | - | 1 - 2 | 96 | 100 | |
More Information
Set Membership: |
|

GLYATL3
Genomic Map