Assay Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145233.1 | NP_001138705.1 | 1 - 2 | 36 | 77 | |
| GenBank mRNA | FJ562213.1 | - | 1 - 2 | 36 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001145233.1 transcripts.
Set Membership: |
|

FFAR3
Genomic Map