Assay Details
| Gene Symbol: ATRAID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105505.1 | NP_001098975.1 | 5 - 6 | 507 | 85 | |
| GenBank mRNA | BC149353.1 | - | 5 - 6 | 507 | 85 | |
| BT029884.1 | - | 5 - 6 | 499 | 85 | ||
| BT029908.1 | - | 5 - 6 | 499 | 85 | ||
More Information
Set Membership: |
|

ATRAID
Genomic Map