Assay Details
| Gene Symbol: HSD11B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123032.1 | NP_001116504.1 | 4 - 5 | 573 | 68 | |
| GenBank mRNA | AF548027.2 | - | 4 - 5 | 573 | 68 | |
More Information
Set Membership: |
|

HSD11B1
Genomic Map