Assay Details
| Gene Symbol: LIMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130751.1 | NP_001124223.1 | 4 - 5 | 431 | 66 | |
| GenBank mRNA | BC105544.1 | - | 4 - 5 | 431 | 66 | |
More Information
Set Membership: |
|

LIMS2
Genomic Map