Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101185.1 | NP_001094655.1 | 2 - 3 | 445 | 81 | |
| NM_001168662.1 | NP_001162133.1 | 1 - 2 | 247 | 81 | ||
| GenBank mRNA | BC149355.1 | - | 1 - 2 | 248 | 81 | |
| EE243393.1 | - | 2 - 3 | 390 | 81 | ||
More Information
Set Membership: |
|

CYP26A1
Genomic Map