Assay Details
| Gene Symbol: TSC22D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103342.1 | NP_001096812.1 | 2 - 3 | 489 | 72 | |
| GenBank mRNA | BC148873.1 | - | 2 - 3 | 489 | 72 | |
More Information
Set Membership: |
|

TSC22D3
Genomic Map