Assay Details
| Gene Symbol: CDYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163785.1 | NP_001157257.1 | 6 - 7 | 1391 | 68 | |
| GenBank mRNA | EF690283.1 | - | 6 - 7 | 1391 | 68 | |
More Information
Set Membership: |
|

CDYL2
Genomic Map