Assay Details
| Gene Symbol: VNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163920.1 | NP_001157392.1 | 3 - 4 | 539 | 97 | |
| GenBank mRNA | BT030550.1 | - | 3 - 4 | 539 | 97 | |
More Information
Set Membership: |
|

VNN2
Genomic Map