Assay Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205486.1 | NP_001192415.1 | 6 - 7 | 916 | 59 | |
| GenBank mRNA | BC142221.1 | - | 6 - 7 | 899 | 59 | |
More Information
Set Membership: |
|

UBE2Z
Genomic Map