Assay Details
| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205573.3 | NP_001192502.2 | 14 - 15 | 1682 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LARP4B| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205573.3 | NP_001192502.2 | 14 - 15 | 1682 | 60 | |
Set Membership: |
|