Assay Details
| Gene Symbol: SLC39A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205630.1 | NP_001192559.1 | 7 - 8 | 1424 | 86 | |
| GenBank mRNA | MK341547.1 | - | 1 - 2 | 48 | 86 | |
More Information
Set Membership: |
|

SLC39A8
Genomic Map