Assay Details
| Gene Symbol: NDFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205720.1 | NP_001192649.1 | 2 - 3 | 523 | 87 | |
| GenBank mRNA | BC105183.1 | - | 2 - 3 | 376 | 87 | |
More Information
Set Membership: |
|

NDFIP2
Genomic Map