Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205724.2 | NP_001192653.1 | 5 - 6 | 890 | 72 | |
| GenBank mRNA | BC151787.1 | - | 5 - 6 | 698 | 72 | |
| EV669788.1 | - | 5 - 6 | 632 | 72 | ||
| EV690720.1 | - | 5 - 6 | 590 | 72 | ||
More Information
Set Membership: |
|

DNASE1L3
Genomic Map