Assay Details
| Gene Symbol: DLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205730.2 | NP_001192659.2 | 12 - 13 | 1724 | 90 | |
| GenBank mRNA | DT900308.1 | - | 5 - 6 | 524 | 90 | |
More Information
Set Membership: |
|

DLAT
Genomic Map