Assay Details
| Gene Symbol: FAM21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205767.2 | NP_001192696.1 | 1 - 2 | 109 | 65 | |
| GenBank mRNA | BC151722.1 | - | 1 - 2 | 110 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151722.1, NM_001205767.2 transcripts.
Set Membership: |
|

FAM21A
Genomic Map