Assay Details
| Gene Symbol: XKRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205914.1 | NP_001192843.1 | 2 - 3 | 607 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XKRX| Gene Symbol: XKRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205914.1 | NP_001192843.1 | 2 - 3 | 607 | 106 | |
Set Membership: |
|