Assay Details
| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205982.1 | NP_001192911.1 | 2 - 3 | 891 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NODAL| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205982.1 | NP_001192911.1 | 2 - 3 | 891 | 100 | |
Set Membership: |
|