Assay Details
| Gene Symbol: SLC35E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192646.2 | NP_001179575.1 | 1 - 2 | 401 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192646.2 transcripts.
Set Membership: |
|

SLC35E2
Genomic Map