Assay Details
| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206276.2 | NP_001193205.1 | 5 - 6 | 712 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WIPF2| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206276.2 | NP_001193205.1 | 5 - 6 | 712 | 85 | |
Set Membership: |
|