Assay Details
| Gene Symbol: CLDND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206357.1 | NP_001193286.1 | 3 - 4 | 462 | 88 | |
| GenBank mRNA | BC122592.1 | - | 3 - 4 | 393 | 88 | |
| DT813987.1 | - | 2 - 3 | 411 | 88 | ||
More Information
Set Membership: |
|

CLDND1
Genomic Map