Assay Details
| Gene Symbol: TMED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206433.1 | NP_001193362.1 | 3 - 4 | 428 | 63 | |
| GenBank mRNA | BC123580.1 | - | 3 - 4 | 429 | 63 | |
More Information
Set Membership: |
|

TMED4
Genomic Map