Assay Details
| Gene Symbol: PSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206476.1 | NP_001193405.1 | 6 - 7 | 795 | 67 | |
| GenBank mRNA | AF474175.1 | - | 3 - 4 | 314 | 67 | |
| BC120106.1 | - | 6 - 7 | 742 | 67 | ||
| BC151754.1 | - | 6 - 7 | 744 | 67 | ||
More Information
Set Membership: |
|

PSIP1
Genomic Map