Assay Details
| Gene Symbol: CD2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206539.1 | NP_001193468.1 | 2 - 3 | 233 | 72 | |
| GenBank mRNA | BC102654.1 | - | 2 - 3 | 233 | 72 | |
More Information
Set Membership: |
|

CD2AP
Genomic Map