Assay Details
| Gene Symbol: LOC534630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206548.1 | NP_001193477.1 | 1 - 2 | 50 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206548.1 transcripts.
Set Membership: |
|

LOC534630
Genomic Map