Assay Details
| Gene Symbol: LOC534630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206548.1 | NP_001193477.1 | 2 - 3 | 167 | 76 | |
| GenBank mRNA | BC134763.1 | - | 2 - 3 | 156 | 76 | |
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256589.1 | NP_001243518.1 | 2 - 3 | 185 | 76 | |
| GenBank mRNA | EV628920.1 | - | 2 - 3 | 189 | 76 | |
More Information
Set Membership: |
|

LOC534630
Genomic Map